Learn More
Pion Inc ACCEPTOR SINK BUFFER 250ML
Catalog No. NC0875680
Encompass
Proprietary acceptor solution for performing double-sink assays. Quantity: 1 bottle; volume: 250 ml per bottle.
Catalog No. NC0875680 Supplier Pion Inc Supplier No. 110139
Proprietary acceptor solution for performing double-sink assays. Quantity: 1 bottle; volume: 250 ml per bottle.