Learn More
Beckman Coulter Access 2
Catalog No. 21280118 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
Catalog No. For Use With (Equipment)
21-280-118 Beckman Coulter Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 21-280-118 Supplier Beckman Coulter Supplier No. 973111
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Access 2 Immunoassay System is a powerful benchtop system that maximizes valuable lab space.
  • Standardized test menus, assay protocols, instrument processes and reference ranges
  • Maximized productivity in a minimal footprint
  • Reliable uptime with fewer emergency service calls
  • Established performance for confidence in patient results
  • Lower operating costs
TRUSTED_SUSTAINABILITY
Specifications
Type Automated Enzyme Immunoassay Analyzer
For Use With (Equipment) Beckman Coulter Analyzer
Throughput 100 Tests/hr.
Capacity 60 Samples