Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access BNP

Catalog No. 21280117 Shop All Beckman Coulter Products
Change view
Click to view available options
Detectable Analytes:
BNP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
21-280-117 BNP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-117 Supplier Beckman Coulter Supplier No. 98200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access BNP assay offers exceptional clinical quality for cardiac testing.

  • B-type natriuretic peptide (BNP) levels are related to the severity of signs and symptoms of heart failure
  • Access BNP is the only BNP assay on automated immunoassay platforms with two risk stratification claims FDA cleared for patients with Heart Failure and patients with Acute Coronary Syndrome
  • Outstanding Analytical Sensitivity: 1 pg/mL
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes BNP
Sensitivity 1 pg/mL
Type Reagent
Disposable Limited-use
For Use With (Equipment) Beckman Coulter Analyzer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.