Learn More
Beckman Coulter Access Cortisol Calibrators S0
Catalog No. 21280141 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
Catalog No. For Use With (Equipment)
21-280-141 Beckman Coulter Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 21-280-141 Supplier Beckman Coulter Supplier No. 33606
Add to Cart
Add to Cart
This item is not returnable. View return policy
Access Cortisol assay offers exceptional clinical quality for adrenal/pituitary testing.
  • Regulates protein, fat, and carbohydrate metabolism; maintains muscle and myocardial integrity, and suppresses inflammatory and allergic activities
  • No extraction for urines required
  • Multiple sample types allowed: serum, plasma (EDTA and heparin) and urine
  • Minimal cross-reactivity with dexamethasone
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator