Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Cortisol Determinations

Catalog No. 21280143 Shop All Beckman Coulter Products
Change view
Click to view available options
Detection Method:
One-step Competitive
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
21-280-143 One-step Competitive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-143 Supplier Beckman Coulter Supplier No. 33600
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Cortisol assay offers exceptional clinical quality for adrenal/pituitary testing.

  • Regulates protein, fat, and carbohydrate metabolism; maintains muscle and myocardial integrity, and suppresses inflammatory and allergic activities
  • No extraction for urines required
  • Multiple sample types allowed: serum, plasma (EDTA and heparin) and urine
  • Minimal cross-reactivity with dexamethasone
TRUSTED_SUSTAINABILITY

Specifications

CE Marker Yes
Clia Complexity High, Moderate
Detectable Analytes Cortisol
Detection Method One-step Competitive
DoA Calibrators No
Sample Type Serum or Plasma, Urine
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Form Liquid Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.