Learn More
Grainger TOUGH GUY Access Door, Steel, 24 in x 24 in, 16 ga, 1 ea
Catalog No. 502789261
Encompass_Preferred
This access door provides secure and clean access to mechanical, electrical, and plumbing systems concealed within walls or ceilings. Designed for drywall installations, the 24 in x 24 in steel unit features a durable piano hinge and a drywall bead flange to ensure a professional, flush finish in commercial and industrial settings.
  • Constructed from 16 ga steel with a white powder coated finish
  • Equipped with a hex head cam latch mechanism
  • Features a recessed panel design
  • Includes a drywall bead flange for seamless integration
  • Suitable for wall or ceiling mounting
  • Uninsulated design
Catalog No. 50-278-9261 Supplier Grainger Supplier No. 16M222
This access door provides secure and clean access to mechanical, electrical, and plumbing systems concealed within walls or ceilings. Designed for drywall installations, the 24 in x 24 in steel unit features a durable piano hinge and a drywall bead flange to ensure a professional, flush finish in commercial and industrial settings.
  • Constructed from 16 ga steel with a white powder coated finish
  • Equipped with a hex head cam latch mechanism
  • Features a recessed panel design
  • Includes a drywall bead flange for seamless integration
  • Suitable for wall or ceiling mounting
  • Uninsulated design