Learn More
Grainger Tough Guy Access Door, 12 in x 12 in, Steel, Drywall Bead Flange, 1 ea
Catalog No. 502790338
Encompass_Preferred
This access door is designed to provide secure, concealed access to plumbing, electrical, and heating systems within walls or ceilings. Constructed from durable 16 ga steel with a white powder-coated finish, it features a recessed panel and a drywall bead flange for seamless integration. The door is equipped with a hex head cam latch mechanism to limit access and can be locked for added security.
  • Features a durable 16 ga steel construction with a white powder-coated finish.
  • Designed for recessed mounting in walls or ceilings.
  • Includes a drywall bead flange for clean, professional installation.
  • Equipped with a hex head cam latch and a piano hinge for smooth, downward operation.
  • Suitable for areas requiring protection or limited access to internal equipment.
  • Includes 8 mounting holes with a 1/4 in. diameter.
Catalog No. 50-279-0338 Supplier Grainger Supplier No. 16M221
This access door is designed to provide secure, concealed access to plumbing, electrical, and heating systems within walls or ceilings. Constructed from durable 16 ga steel with a white powder-coated finish, it features a recessed panel and a drywall bead flange for seamless integration. The door is equipped with a hex head cam latch mechanism to limit access and can be locked for added security.
  • Features a durable 16 ga steel construction with a white powder-coated finish.
  • Designed for recessed mounting in walls or ceilings.
  • Includes a drywall bead flange for clean, professional installation.
  • Equipped with a hex head cam latch and a piano hinge for smooth, downward operation.
  • Suitable for areas requiring protection or limited access to internal equipment.
  • Includes 8 mounting holes with a 1/4 in. diameter.