Learn More
Grainger TOUGH GUY Access Door, 12 in. x 12 in., Steel, 16 ga., 1 ea.
Catalog No. 502789246
Encompass_Preferred
This access door provides a secure and aesthetic solution for concealing plumbing, heating, and electrical components within wall or ceiling installations. Constructed from durable 16 ga steel with a white powder-coated finish, the door features a flush mount design that integrates seamlessly into surfaces.
  • Concealed hinge provides a clean, professional appearance
  • Screwdriver operated cam latch ensures secure closure
  • Designed for wall or ceiling mounting
  • Uninsulated construction
  • Eight 1/4 in. mounting holes for easy installation
  • 12 in. x 12 in. nominal door size with a 12-1/4 in. x 12-1/4 in. rough opening requirement
Catalog No. 50-278-9246 Supplier Grainger Supplier No. 16M210
This access door provides a secure and aesthetic solution for concealing plumbing, heating, and electrical components within wall or ceiling installations. Constructed from durable 16 ga steel with a white powder-coated finish, the door features a flush mount design that integrates seamlessly into surfaces.
  • Concealed hinge provides a clean, professional appearance
  • Screwdriver operated cam latch ensures secure closure
  • Designed for wall or ceiling mounting
  • Uninsulated construction
  • Eight 1/4 in. mounting holes for easy installation
  • 12 in. x 12 in. nominal door size with a 12-1/4 in. x 12-1/4 in. rough opening requirement