Learn More
Grainger Tough Guy Access Door, Steel, 14 in. x 14 in., Concealed Hinge, 1 ea.
Catalog No. 502789331
Encompass_Preferred
This access door provides secure and aesthetic entry to plumbing, heating systems, and electrical equipment concealed within walls or ceilings. Constructed from 16 ga steel with a white powder-coated finish, it is designed for flush mounting in a variety of service applications.
  • Flush mount design for a clean, professional aesthetic
  • Hidden flange for improved appearance
  • Includes a screwdriver cam latch
  • Concealed hinge allows for a downward door swing
  • Constructed from durable 16 ga steel
  • Suitable for installation in wall or ceiling applications
  • Requires 14 1/4 in. x 14 1/4 in. rough opening
Catalog No. 50-278-9331 Supplier Grainger Supplier No. 16M211
This access door provides secure and aesthetic entry to plumbing, heating systems, and electrical equipment concealed within walls or ceilings. Constructed from 16 ga steel with a white powder-coated finish, it is designed for flush mounting in a variety of service applications.
  • Flush mount design for a clean, professional aesthetic
  • Hidden flange for improved appearance
  • Includes a screwdriver cam latch
  • Concealed hinge allows for a downward door swing
  • Constructed from durable 16 ga steel
  • Suitable for installation in wall or ceiling applications
  • Requires 14 1/4 in. x 14 1/4 in. rough opening