Learn More
Grainger TOUGH GUY Access Door, 16 in. x 16 in., Steel, Concealed Hinge, 1 ea.
Catalog No. 502789332
Encompass_Preferred
This access door is designed to provide secure, concealed access to plumbing, heating systems, and electrical equipment within walls or ceilings. Constructed from 16 ga steel with a white powder-coated finish, it ensures durability and a clean aesthetic. The door features a concealed hinge and a screwdriver-operated cam latch to maintain a flush profile while limiting unauthorized access to sealed-off areas.
  • Concealed hinge for a clean, flush appearance
  • Screwdriver-operated cam latch included for secure closure
  • Constructed from 16 ga steel with a white powder-coated finish
  • Suitable for both wall and ceiling installations
  • Uninsulated design
  • Features 8 mounting holes with 1/4 in. diameter
  • Required rough opening size of 16-1/4 in. x 16-1/4 in.
Catalog No. 50-278-9332 Supplier Grainger Supplier No. 16M212
This access door is designed to provide secure, concealed access to plumbing, heating systems, and electrical equipment within walls or ceilings. Constructed from 16 ga steel with a white powder-coated finish, it ensures durability and a clean aesthetic. The door features a concealed hinge and a screwdriver-operated cam latch to maintain a flush profile while limiting unauthorized access to sealed-off areas.
  • Concealed hinge for a clean, flush appearance
  • Screwdriver-operated cam latch included for secure closure
  • Constructed from 16 ga steel with a white powder-coated finish
  • Suitable for both wall and ceiling installations
  • Uninsulated design
  • Features 8 mounting holes with 1/4 in. diameter
  • Required rough opening size of 16-1/4 in. x 16-1/4 in.