Learn More
Grainger Tough Guy Access Door, 18 in. x 18 in., Steel, Concealed Hinge, 1 ea
Catalog No. 502790677
Encompass_Preferred
This access door provides a secure solution for concealing and protecting plumbing, heating systems, and electrical equipment behind wall or ceiling surfaces. Designed for flush mounting, the unit features a durable 16 ga steel construction with a white powder-coated finish for long-lasting performance.
  • 18 in. x 18 in. dimensions
  • Concealed hinge with downward swing
  • Screwdriver operated cam latch
  • Uninsulated design
  • Hidden flange for aesthetic appeal
  • Suitable for wall or ceiling installation
  • Includes 8 mounting holes with 1/4 in. diameter
  • 18-1/4 in. x 18-1/4 in. required rough opening
Catalog No. 50-279-0677 Supplier Grainger Supplier No. 16M213
This access door provides a secure solution for concealing and protecting plumbing, heating systems, and electrical equipment behind wall or ceiling surfaces. Designed for flush mounting, the unit features a durable 16 ga steel construction with a white powder-coated finish for long-lasting performance.
  • 18 in. x 18 in. dimensions
  • Concealed hinge with downward swing
  • Screwdriver operated cam latch
  • Uninsulated design
  • Hidden flange for aesthetic appeal
  • Suitable for wall or ceiling installation
  • Includes 8 mounting holes with 1/4 in. diameter
  • 18-1/4 in. x 18-1/4 in. required rough opening