Learn More
Grainger Tough Guy Access Door, Fire Rated, Steel, 8 in. x 8 in., 1 ea.
Catalog No. 502789298
Encompass_Preferred
This fire-rated access door is designed for installation in walls or ceilings to provide secure, flame-resistant access to building components. The unit features an insulated steel construction with a white powder-coated finish and is engineered for flush mounting.
  • Fire rating of 1-1/2 hr in walls, 3 hr in noncombustible ceilings, and 1 hr in combustible ceilings.
  • Includes a ring operated slam latch and self-latching tool key for secure closure.
  • Constructed with a 16 ga. cold rolled steel frame and a 20 ga. steel door.
  • Features a piano hinge for smooth operation with a downward door swing.
  • TAA compliant and certified to meet multiple fire safety standards, including UL 10B and NFPA 252.
Catalog No. 50-278-9298 Supplier Grainger Supplier No. 16M205
This fire-rated access door is designed for installation in walls or ceilings to provide secure, flame-resistant access to building components. The unit features an insulated steel construction with a white powder-coated finish and is engineered for flush mounting.
  • Fire rating of 1-1/2 hr in walls, 3 hr in noncombustible ceilings, and 1 hr in combustible ceilings.
  • Includes a ring operated slam latch and self-latching tool key for secure closure.
  • Constructed with a 16 ga. cold rolled steel frame and a 20 ga. steel door.
  • Features a piano hinge for smooth operation with a downward door swing.
  • TAA compliant and certified to meet multiple fire safety standards, including UL 10B and NFPA 252.