Learn More
Grainger TOUGH GUY Access Door, 8 in x 8 in, Steel, Piano Hinge, Exposed Flange, 1 ea
Catalog No. 502789373
Encompass_Preferred
This steel access door provides a secure solution for concealing and protecting plumbing, heating systems, and electrical equipment behind wall installations. The flush-mounted, uninsulated door features a durable powder-coated finish and is designed for use on vertical fire-rated surfaces where temperature rise protection is not required.
  • 60 min fire rating
  • Piano hinge with downward door swing
  • Built-in keyed lock for restricted access
  • Includes self-latching tool key and ring-operated slam latch
  • 16 ga steel construction
  • 8 in x 8 in dimensions with exposed flange
  • Meets multiple fire safety standards including NFPA 252 and UL 10B
Catalog No. 50-278-9373 Supplier Grainger Supplier No. 16M215
This steel access door provides a secure solution for concealing and protecting plumbing, heating systems, and electrical equipment behind wall installations. The flush-mounted, uninsulated door features a durable powder-coated finish and is designed for use on vertical fire-rated surfaces where temperature rise protection is not required.
  • 60 min fire rating
  • Piano hinge with downward door swing
  • Built-in keyed lock for restricted access
  • Includes self-latching tool key and ring-operated slam latch
  • 16 ga steel construction
  • 8 in x 8 in dimensions with exposed flange
  • Meets multiple fire safety standards including NFPA 252 and UL 10B