Learn More
Grainger Tough Guy Fire Rated Access Door, Steel, 16 in x 16 in, Uninsulated, 1 ea
Catalog No. 502790601
Encompass_Preferred
This fire-rated, uninsulated access door is designed for installation on vertical surfaces where temperature rise protection is not required. Constructed from steel with a white powder-coated finish, it is engineered for flush wall mounting and provides a reliable fire barrier.
  • 1-1/2 hr. B-label fire rating for 2-hr. rated walls
  • Self-latching design with tool key and ring-operated slam latch
  • Piano hinge style with downward door swing
  • Constructed from 16 ga. steel
  • Certified to CAN/ULC S104 (2010) standards
  • Includes 8 mounting holes with 1/4 in. diameter
Catalog No. 50-279-0601 Supplier Grainger Supplier No. 16M217
This fire-rated, uninsulated access door is designed for installation on vertical surfaces where temperature rise protection is not required. Constructed from steel with a white powder-coated finish, it is engineered for flush wall mounting and provides a reliable fire barrier.
  • 1-1/2 hr. B-label fire rating for 2-hr. rated walls
  • Self-latching design with tool key and ring-operated slam latch
  • Piano hinge style with downward door swing
  • Constructed from 16 ga. steel
  • Certified to CAN/ULC S104 (2010) standards
  • Includes 8 mounting holes with 1/4 in. diameter