Learn More
Grainger Tough Guy Access Door, 18 in. x 18 in., Steel, 16 ga., Powder Coated, 1 ea.
Catalog No. 502790788
Encompass_Preferred
This access door is designed to provide secure, concealed access to plumbing, heating, and electrical systems within walls. Constructed from durable 16 ga steel with a white powder-coated finish, this door is built for long-term use and includes a built-in keyed lock to limit access to sensitive areas.
  • 60 min fire rating
  • Piano hinge with downward door swing
  • Exposed flange for flush mounting
  • Self-latching design with tool key and ring-operated slam latch
  • Uninsulated construction
  • Flame and fire resistant properties
  • Designed for wall installation with 8 mounting holes
Catalog No. 50-279-0788 Supplier Grainger Supplier No. 16M218
This access door is designed to provide secure, concealed access to plumbing, heating, and electrical systems within walls. Constructed from durable 16 ga steel with a white powder-coated finish, this door is built for long-term use and includes a built-in keyed lock to limit access to sensitive areas.
  • 60 min fire rating
  • Piano hinge with downward door swing
  • Exposed flange for flush mounting
  • Self-latching design with tool key and ring-operated slam latch
  • Uninsulated construction
  • Flame and fire resistant properties
  • Designed for wall installation with 8 mounting holes