Learn More
Grainger Tough Guy Access Door, Steel, Flush Mount, 18 in x 18 in, 1 ea
Catalog No. 502789245
Encompass_Preferred
This flush-mount access door is constructed from 16 ga steel and finished with a durable powder coating, providing a secure and accessible panel for wall or ceiling installations. The unit is designed for uninsulated applications and features a downward door swing for ease of use in restricted spaces.
  • Features a key-operated cylinder cam latch for secure closure.
  • Equipped with a hidden pin hinge design for a clean, streamlined appearance.
  • Suitable for wall or ceiling mounting.
  • Includes eight 1/4 in. diameter mounting holes for quick and easy installation.
  • Designed with an exposed flange for a flush finish.
  • Rough opening size is 18 1/4 in. x 18 1/4 in.
Catalog No. 50-278-9245 Supplier Grainger Supplier No. 16M208
This flush-mount access door is constructed from 16 ga steel and finished with a durable powder coating, providing a secure and accessible panel for wall or ceiling installations. The unit is designed for uninsulated applications and features a downward door swing for ease of use in restricted spaces.
  • Features a key-operated cylinder cam latch for secure closure.
  • Equipped with a hidden pin hinge design for a clean, streamlined appearance.
  • Suitable for wall or ceiling mounting.
  • Includes eight 1/4 in. diameter mounting holes for quick and easy installation.
  • Designed with an exposed flange for a flush finish.
  • Rough opening size is 18 1/4 in. x 18 1/4 in.