Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Ferritin Determinations

Catalog No. 21280145 Shop All Beckman Coulter Products
Change view
Click to view available options
Detection Method:
Two-site Sandwich
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
21-280-145 Two-site Sandwich
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-145 Supplier Beckman Coulter Supplier No. 33020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Ferritin assay offers exceptional clinical quality for anemia testing.

  • Aids in the diagnosis of iron deficiency anemia (IDA)
  • Standardized to WHO 3rd International Standard 94/572
  • No hook effect up to 40,000 ng/mL
TRUSTED_SUSTAINABILITY

Specifications

CE Marker Yes
Clia Complexity High, Moderate
Detectable Analytes Ferritin
Detection Method Two-site Sandwich
DoA Calibrators No
Sample Type Serum or Plasma
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.