Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Folate Calibrator S0

Catalog No. 21280107 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-107 Beckman Coulter Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-107 Supplier Beckman Coulter Supplier No. A99250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Folate assay offers exceptional clinical quality for anemia testing.

  • Folate levels in serum, lithium heparin plasma, and red blood cells are used to assess folate status. The serum folate levels is an indicator of recent folate intake. A low RBC folate value can indicate a prolonged folate deficiency.
  • One calibration curve for both serum and RBC applications
  • Folate calibrator accuracy within ± 10% of the folate WHO 03/178
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Folate
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Disposable Limited-use
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.