Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Free T4 Calibrators S0-S5

Catalog No. 21280137 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-137 Beckman Coulter Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-137 Supplier Beckman Coulter Supplier No. 33885
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Free T4 assay offers exceptional clinical quality for thyroid testing.

  • Regulates normal growth and maintenance
  • Two-step format minimizes interferences from HAMA, antithyroid antibodies and thyroxine-binding molecules found in human serum and plasma
  • Optimized formulation minimizes impact to bound/unbound equilibrium
  • Reference interval established using NACB guidelines
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Thyroxine Free T4
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.