Learn More
Binder Inc Access port 30 mm right CB
SDP
Supplier:  Binder Inc 86120019
Encompass_Preferred
Access port with silicone plug - 30 mm, right - optional for BINDER CO2 incubator
Catalog No. NC2064835