Description
Provide a point of entry for probes and monitoring devices in environmental chambers and incubators with Access Ports.
Specifications
Specifications
| Description | Access Port |
| For Use With (Equipment) | 3962 |
| Lighting | LED |
| Type | Access Port |
| Installation | Factory, customer specified location |
| Diameter (Metric) Inside | 61 mm |