Learn More
BINDER™ Top Access Port (50 mm)
SDP
Catalog No. 08111447 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-111-447 Steel
1 options
Catalog No. 08-111-447 Supplier BINDER™ Supplier No. 80120049
Add to Cart
Add to Cart
For Use With: BINDER units series BD
  • For use on top
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 50 mm, top
For Use With (Equipment) BINDER Chambers Series BD
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 50 mm