Learn More
BINDER™ Right Access Port (50 mm)
SDP
Catalog No. 08111461 Shop All BINDER Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
50 mm
Catalog No. Diameter (Metric) Inside
08-111-461 50 mm
1 options
Catalog No. 08-111-461 Supplier BINDER™ Supplier No. 80120541
Add to Cart
Add to Cart
Material: Silicone; For Use With: BINDER units ED 23, ED 400
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 50 mm, right
For Use With (Equipment) BINDER Chambers ED 23, ED 400
Material Silicone
Type Plug
Includes Silicone plug
Diameter (Metric) Inside 50 mm