Learn More
BINDER™ Access Port with Silicone Plug for CBF 170 CO2 Incubators
SDP
Catalog No. 5000217497 Shop All BINDER Inc. Products
Click to view available options
Diameter (Metric):
30 mm
Catalog No. Diameter (Metric)
50-002-17497 30 mm
1 options
Catalog No. 50-002-17497 Supplier BINDER™ Supplier No. 80121392
This Access Port with Silicone Plug is designed for use with BINDER™ CBF 170 CO2 Incubators with Hot Air Sterilization and Humidity Regulation.
  • Access port with silicone plug
  • 30 mm inner diameter
  • Right position
Specifications
Description Right Position
Model BINDER™ CBF 170 CO2 Incubators with Hot Air Sterilization and Humidity Regulation
Diameter (Metric) 30 mm