Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Access Sensitive Estradiol Assay

Catalog No. 01912807 Shop All Beckman Coulter Products
Change view
Click to view available options
No. of Reactions:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
01-912-807 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-912-807 Supplier Beckman Coulter™ Supplier No. B84493
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sensitive estradiol assay for use with Beckman Coulter Access Immunoassay Systems.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Serum, Plasma
For Use With (Equipment) Beckman Coulter Analyzers
No. of Reactions 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.