Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access System Check Solution

Catalog No. 21280121 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-280-121 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-280-121 Supplier Beckman Coulter Supplier No. 81910
Add to Cart
Add to Cart

All Access Immunoassay systems feature standardized reagent and assay protocols that streamline inventory management tasks for labor-cost savings, while providing consistent results.

  • Standardized test menus, assay protocols, instrument processes and reference ranges
  • Maximized productivity in a minimal footprint
  • Reliable uptime with fewer emergency service calls
  • Established performance for confidence in patient results
  • Lower operating costs
TRUSTED_SUSTAINABILITY

Specifications

Type Check Solution
For Use With (Equipment) Beckman Coulter Analyzer
Packaging Type 4 mL Bottles
Quantity 6/Pk.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.