Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Total T4 Determinations

Catalog No. 21280140 Shop All Beckman Coulter Products
Change view
Click to view available options
Detectable Analytes:
Thyroxine (T4 Total)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
21-280-140 Thyroxine (T4 Total)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-140 Supplier Beckman Coulter Supplier No. 33800
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Total T4 assay offers exceptional clinical quality for thyroid testing.

  • Measures total amount of free and bound thyroxine in circulation
  • Integrity; suppresses inflammatory and allergic activities
  • Broad dynamic range
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Thyroxine (T4 Total)
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.