Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access TSH Calibrators S0-S5

Catalog No. 21280093 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-093 Beckman Coulter Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-093 Supplier Beckman Coulter Supplier No. B63285
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access TSH assay offers exceptional clinical quality for thyroid testing.

  • Stimulates the thyroid to make T3 and T4
  • Can discriminate between hyperthyroid and euthyroid subjects
  • 100-test reagent pack
  • Traceable to the WHO 3rd International Standard 81/565 for human TSH
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Thyroid Stimulating Hormone (TSH)
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.