Learn More
Beckman Coulter Access TSH Determinations
Catalog No. 21280094 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
200/Pk.
Catalog No. Quantity
21-280-094 200/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-094 Supplier Beckman Coulter Supplier No. B63284
Add to Cart
Add to Cart
This item is not returnable. View return policy
Access TSH assay offers exceptional clinical quality for thyroid testing.
  • Stimulates the thyroid to make T3 and T4
  • Can discriminate between hyperthyroid and euthyroid subjects
  • 100-test reagent pack
  • Traceable to the WHO 3rd International Standard 81/565 for human TSH
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Thyroid Stimulating Hormone (TSH)
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Quantity 200/Pk.