Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Vitamin B12 Calibrators

Catalog No. 21280147 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
DxI 9000/Beckman Coulter Analyzers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-147 DxI 9000/Beckman Coulter Analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-147 Supplier Beckman Coulter Supplier No. 33005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access B12 assay offers exceptional clinical quality for anemia testing.

  • Aids in the detection of vitamin B12 deficiencies that can lead to megaloblastic anemia and severe neurological issues
  • Automated pretreatment online
  • Direct testing from the primary tube
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0 to 3%
Detectable Analytes Hemoglobin, Bilirubin, Lipemia
Format Solution
For Use With (Equipment) DxI 9000/Beckman Coulter Analyzers
Sample Type Serum, Plasma
Type Calibrator
Stability 14 Days
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.