Description
Includes
TMT10plex Isobaric Label Reagent Set, 3 x 0.8mg, Pierce Peptide Desalting Columns, High Select Top14 Abundant Protein Depletion Mini Spin Columns
- Thermo Scientific High Select Top14 Abundant Protein Depletion Spin Columns decreases the abundant protein and antibody components of human plasma samples, helping prepare the samples for mass spectrometry
- High Select Top14 Abundant Protein Depletion Resin can deplete greater than 95% of the 14 most abundant proteins in serum using highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum and plasma
- Tandem Mass Tag™ (TMT™) 10-plex reagent set contains ten different isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm and a mass reporter. The reagent set enables up to ten different peptide samples prepared from cells or tissues to be labeled in parallel and then combined for analysis
Specifications
Specifications
| Content And Storage | TMT10plex Isobaric Label Reagent Set, 3 x 0.8mg, Pierce Peptide Desalting Columns, High Select Top14 Abundant Protein Depletion Mini Spin Columns. See instructions for individual components |
| Quantity | 1 Bundle |