Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Catalog No. CIC0001448 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
PT1000 External probe 0-400C
Transparent shield for 10x10
Transparent shield for 4x4
Transparent shield for 7x7
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
CIC0001448 PT1000 External probe 0-400C
CIC0001444 Transparent shield for 4x4
CIC0001445 Transparent shield for 7x7
CIC0001446 Transparent shield for 10x10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CIC0001448 Supplier Thermo Scientific™ Supplier No. CIC0001448
  • $137.00
    $127.65 / Each of 1
    Save $9.35
    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Get precise stirring control for your routine protocols.
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Display LED
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Type PT1000 External probe 0-400C