Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Catalog No. p-8444000
Change view
Click to view available options
Type:
PT1000 External probe 0-400C
Transparent shield for 10x10
Transparent shield for 4x4
Transparent shield for 7x7
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
CIC0001448 PT1000 External probe 0-400C
CIC0001444 Transparent shield for 4x4
CIC0001445 Transparent shield for 7x7
CIC0001446 Transparent shield for 10x10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CIC0001448 Supplier Thermo Scientific™ Supplier No. CIC0001448
Only null left
Add to Cart
Add to Cart
Get precise stirring control for your routine protocols.
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Display LED
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Type PT1000 External probe 0-400C