Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ HI-200 Series Hybridization Incubators
Catalog No. p-200004266
Change view
Click to view available options
Type:
Tube Rack Holder
Catalog No. Type
13-245-99C Tube Rack Holder
1 options
Catalog No. 13-245-99C Supplier Cole-Parmer™ Supplier No. 3945527
Complement your hybridization incubation systems
  • Expand the capabilities of your Cole Parmer HI-200 Series Hybridization Incubators with our collection of optional accessories
  • This collection includes a selection of rocking platforms, hybridization tubes of various sizes, and tube holders with clips
Specifications
For Use With (Equipment) Hybridization incubator
Holds Up to 3 Large and 3 Small Tubes
Material Plastic
Type Tube Rack Holder
Includes Hybridization Tube Rack Holder
Length (Metric) 80 mm