Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Catalog No. 88861121 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
1.5/2.0 mL Tubes
10-15 mL and 5-7 mL Tubes
14 mm dia. Culture Tubes
15 mL Tubes
18.5 mm dia. Culture Tubes
50 mL Tubes
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
88-861-121 1.5/2.0 mL Tubes
88-861-122 15 mL Tubes
88-861-123 50 mL Tubes
88-861-124 10-15 mL and 5-7 mL Tubes
88-861-125 14 mm dia. Culture Tubes
88-861-126 18.5 mm dia. Culture Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 88-861-121
Add to Cart
Add to Cart
Optional accessories to enhance the Fisherbrand™Multi-Purpose Tube Rotator
Multi-purpose tube rotator offers flexibility with multiple carousel options
  • Tube rotator
TRUSTED_SUSTAINABILITY
Specifications
Holds 1.5/2.0 mL Tubes
Product Type Tube Carousel
No. of Positions 50