Learn More
Agilent BioTek Accessories For MultiFlo Microplate Dispenser - Manifolds
GSA_VAGREENER_CHOICE
Catalog No. 11120112 Shop All Agilent Technologies Products
Change view
Click to view available options
Plate Type:
24-well
48-well
6-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Type
11-120-112 24-well
11-120-111 6-well
11-120-113 48-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-120-112 Supplier Agilent BioTek Supplier No. 1260535S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accessories for MultiFlo Microplate Dispenser
  • Syringe manifold
  • Available with 6, 24, or 48-well plates
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Plate Type 24-well