Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Accessories for G20 Titrator Models

Catalog No. 01910713 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Cable
Electrode
Screw
Sleeve
Temperature Sensor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-910-713 Cable
01-910-719 Electrode
01-910-720 Temperature Sensor
01-910-714 Sleeve
01-910-704 Screw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-910-713 Supplier Mettler Toledo™ Supplier No. 89601
Add to Cart
Add to Cart

For use with G20 Titrator models.

  • SC-LEMO-60
  • Length: 70cm
TRUSTED_SUSTAINABILITY

Specifications

Type Cable
Length (English) 27.6 in.
Length (Metric) 700 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.