Learn More
Mettler Toledo™ Accessories for G20 Titrator Models
GSA_VA
Catalog No. 01910719 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Cable
Electrode
Screw
Sleeve
Temperature Sensor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-910-719 Electrode
01-910-713 Cable
01-910-720 Temperature Sensor
01-910-714 Sleeve
01-910-704 Screw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-910-719 Supplier Mettler Toledo™ Supplier No. 51109300
Add to Cart
Edge
Add to Cart
Request a Quote
For use with G20 Titrator models.
  • Phototrode with 5 selectable wave lengths 520nm, 555nm, 590nm, 620nm, 660nm
  • Includes LEMO connection cable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode
Includes LEMO connection cable, 5 selectable wavelength