Learn More
HemoCue America Accessories for HemoCue™ 201 DM Analyzers
Catalog No. 22601029
Change view
Click to view available options
Type:
DM Battery
PC Software
USB Cable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-601-029 PC Software
22-601-028 DM Battery
22-601-025 USB Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-601-029 Supplier HemoCue America Supplier No. 131062V3.2
Add to Cart
Add to Cart
For use with 201 DM Analyzers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HemoCue™ 201 DM analyzer
Type PC Software
Includes 1 CD Rom, 1 license per device
Intended Use Software generates patient and QC reports and allows downloading of the data, it also allows downloading of instrument configurations, changing of operator lists, control batches etc
Quantity Each