Learn More
Fisherbrand™ Accessories for Incubating Mini Shaker
Catalog No. 88861144 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Centrifuge Tube Rack
Flask Clamp
Sticky Mat
Holds:
100, 125 mL Flasks
25 mL Flasks
250 mL Flasks
50 mL Centrifuge Tubes
50 mL Flasks
N/A
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
88-861-144 Sticky Mat N/A
88-861-135 Flask Clamp 25 mL Flasks
88-861-138 Flask Clamp 250 mL Flasks
88-861-136 Flask Clamp 50 mL Flasks
88-861-137 Flask Clamp 100, 125 mL Flasks
88-861-103 Centrifuge Tube Rack 50 mL Centrifuge Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 88-861-144
Get an Accessory with Shaker Purchase
View all promotions
Add to Cart
Add to Cart
Accessories required for the use of Fisher Scientific™ Incubating Mini Shaker
Accessories for incubating mini shaker
  • Increase performance and flexibility
TRUSTED_SUSTAINABILITY
Specifications
Holds N/A
Product Type Sticky Mat
Dimensions (L x W) 140 x 140 mm
No. of Positions N/A