Learn More
Marketlab, Inc. Accessories for Insight™ Modular Cart
Description
- Sharps Bracket Special for 2 gal./8 QT
- Ideal for customizing your Insight carts to meet your specific needs
- Accessory Bracket (60873) is compatible with Insight Undercounter Storage Cart (19689), Insight Storage Carts (43350 and 18087), and Insight Phlebotomy Carts (21491 and 138760)
- 1-year limited warranty
Specifications
Specifications
| Depth (English) | 6 in. |
| Width (English) | 12 in. |
| Height (English) | 9 in. |
| Product Type | Sharps Bracket |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.