Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Isotemp™ Support Stand
GSA_VA

Catalog No. 15497017 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
15-497-017 Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-497-017 Supplier Thermo Scientific™ Supplier No. 190648
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Material: Steel; For Use With: Napco 8000 Series CO2 Incubators

  • For Isotemp CO2 Incubators 13-255-25, -26, -27, -28
  • For Napco 8000 Series CO2 Incubators 15-497-001 through -012

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Napco 8000 Series CO2 Incubators
Lighting LED
Material Steel
Type Stand
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.