Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Masterflex™ Accessories for Peristaltic Pump Systems

Catalog No. 11747360
Change view
Click to view available options
Control Type:
Remote I/O
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Control Type
11-747-360 Remote I/O
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-747-360 Supplier Masterflex™ Supplier No. MFLX0752395
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Upgrade pumping and dispensing capabilities with Accessories for Masterflex™ Peristaltic Pump Systems.

  • Accessory items for the Masterflex™ Peristaltic Pump Systems

Warranty and Services

2 Years

Specifications

Certifications/Compliance ISO 9002, CE
Color Gray
Control Type Remote I/O
For Use With (Application) Fluid handling
Includes Masterflex Cable Assembly for Remote Control, 25 ft. (7.6 m) cable with stripped-wire ends, DB25(M) connector
Speed Control Analog
Overall Length (English) 25 ft.
Overall Length (Metric) 7.6 m
Weight (English) 1.26 lb.
Weight (Metric) 571.53 g
For Use With (Equipment) Masterflex drives with DB-25 connection and remote control capability
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.