Learn More
MilliporeSigma™ Accessories to Milli-Q™ IQ 7000 Ultrapure Water System
Catalog No. ZFC0NDCA1 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Feed Water Conductivity Cell
Foot Pedal
Water Sensor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
ZFC0NDCA1 Feed Water Conductivity Cell
ZMQSFTSA1 Foot Pedal
ZWATSENA1 Water Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ZFC0NDCA1 Supplier MilliporeSigma™ Supplier No. ZFC0NDCA1
Add to Cart
Add to Cart
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories -- in both validated and non-validated environments.
Water purification accessories.
Feed Water Conductivity Cell :
Measures the conductivity of water feeding
Foot Pedal: Hands-free dispensing accessory
Tank Level Adapter:  For pure feed water stored in lab water storage tank 
Water Sensor: Detects water spillage and automatically closes the Milli-Q™ inlet solenoid valve to protect the surroundings from water damage; 120V/60Hz
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
TRUSTED_SUSTAINABILITY
Specifications
Type Feed Water Conductivity Cell
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System