Learn More
MilliporeSigma™ Accessories to Milli-Q™ IQ 7000 Ultrapure Water System
GSA_VA
Catalog No. ZWATSENA1 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Feed Water Conductivity Cell
Foot Pedal
Water Sensor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
ZWATSENA1 Water Sensor
ZFC0NDCA1 Feed Water Conductivity Cell
ZMQSFTSA1 Foot Pedal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ZWATSENA1 Supplier MilliporeSigma™ Supplier No. ZWATSENA1
Add to Cart
Add to Cart
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories -- in both validated and non-validated environments.
Water purification accessories.
Feed Water Conductivity Cell :
Measures the conductivity of water feeding
Foot Pedal: Hands-free dispensing accessory
Tank Level Adapter:  For pure feed water stored in lab water storage tank 
Water Sensor: Detects water spillage and automatically closes the Milli-Q™ inlet solenoid valve to protect the surroundings from water damage; 120V/60Hz
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Water Sensor
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System