Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Shop All Thermo Scientific ProductsDescription
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Features:
- Accessories for AQUAfast AQ3010 Turbidity Meter
Specifications
Specifications
| Type | Vial Cleaning Brushes |
| For Use With (Equipment) | AQUAfast AQ3010 Turbidity Meter |
| Description | Vial Cleaning Brushes, 5 Pack |