Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Catalog No. p-7114726 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Silicone Oil
Vial Cleaning Brushes
Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-644-500 Vial Cleaning Brushes
13-644-502 Silicone Oil
13-644-503 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-644-500 Supplier Thermo Scientific™ Supplier No. AC3CBR
Add to Cart
Add to Cart
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Features:
  • Accessories for AQUAfast AQ3010 Turbidity Meter
TRUSTED_SUSTAINABILITY
Specifications
Type Vial Cleaning Brushes
For Use With (Equipment) AQUAfast AQ3010 Turbidity Meter
Description Vial Cleaning Brushes, 5 Pack