Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Heracell™ VIOS 160i/Thermo Scientific™ Forma™ Steri-Cycle™
Shop All Thermo Scientific ProductsSpecifications
Specifications
| For Use With (Equipment) | Thermo Scientific Heracell VIOS 160i AND Thermo Scientific Forma Steri-Cycle i160 Incubators |
| Type | Stacking Adapter |
| For Use With (Application) | Configured to stack a 160 L on top of 184 L incubator |