Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Catalog No. 13872273
Change view
Click to view available options
Type:
Cage
Extension Bridge
Female M16x1 Adapter (NPTF)
Fluid Displacement Block
Lifting Platform
Male M16x1 Adapter (NPTF)
Tap Water Cooling Coil
Trolley with Castors
For Use With (Equipment):
AC150 or AC200 controller with S3
Cold/Heated Bath Circulators
For A10
For A10B
For A25 or A40
For A28
For A40
For A5B
For S15 Heated Bath Circulator
For S15, S21, S30, S45
S49 Liftplate
SC100 or SC150 Immersion Circulator
SC100/SC150 Circulator with S3
SC100/SC150 Circulator with S6T
SC150L Circulator with S7
SC150L Immersion Circulator
SC150L Immersion Circulator+clamp
SC150L with S13, S15, S30, S45, S49
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
13-872-273 Extension Bridge S49 Liftplate
13-265-112 Lifting Platform For S15 Heated Bath Circulator
13-265-115 Tap Water Cooling Coil SC100 or SC150 Immersion Circulator
13-265-116 Tap Water Cooling Coil SC150L with S13, S15, S30, S45, S49
13-265-117 Tap Water Cooling Coil SC150L Immersion Circulator+clamp
13-873-884 Female M16x1 Adapter (NPTF) Cold/Heated Bath Circulators
13-874-771 Fluid Displacement Block For A10
13-265-104 Trolley with Castors For A40
13-872-272 Extension Bridge For S15, S21, S30, S45
13-265-106 Cage SC100 or SC150 Immersion Circulator
13-265-107 Cage SC150L Immersion Circulator
13-265-120 Tap Water Cooling Coil SC100/SC150 Circulator with S6T
13-265-121 Tap Water Cooling Coil SC100/SC150 Circulator with S3
13-265-122 Tap Water Cooling Coil SC150L Circulator with S7
13-265-123 Tap Water Cooling Coil AC150 or AC200 controller with S3
13-874-777 Fluid Displacement Block For A28
13-874-778 Lifting Platform For A5B
13-874-779 Lifting Platform For A10B
13-873-885 Male M16x1 Adapter (NPTF) Cold/Heated Bath Circulators
13-874-776 Fluid Displacement Block For A25 or A40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 13-872-273 Supplier Thermo Scientific™ Supplier No. 1600009
Add to Cart
Add to Cart
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S49 Liftplate
Type Extension Bridge