Learn More
Thermo Scientific™ Accessories for TSG Series Countertop Refrigerators
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ TSG Series Countertop Refrigerators have several accessory options available. E-lock adapter kits are available for Pyxis™ and Omnicell™ systems, and a 8.5 in. rollerbase makes moving the TSG205SA from room-to-room easy and convenient.
- OF02 is compatible with Omnicellock™ systems
- PA02 is compatible with Pyxis™lock systems
- RB02 is an 8.5" roller base for the TSG205SA
Specifications
Specifications
| Type | Pyxis Lock Adapter |
| Style | Countertop |
| For Use With (Equipment) | TSG Series Countertop Refrigerator |
| Access Security | Pyxis Lock Adapter |